Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP62CL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012107591.3 | XP_011962981.1 | 6 - 7 | 717 | 113 | |
XM_012107592.3 | XP_011962982.1 | 6 - 7 | 720 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |