Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959276.1 | XP_027815077.1 | 3 - 4 | 654 | 85 | |
XM_027959277.1 | XP_027815078.1 | 3 - 4 | 2726 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |