Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004003864.4 | XP_004003913.2 | 2 - 3 | 595 | 88 | |
XM_015092607.2 | XP_014948093.2 | 2 - 3 | 201 | 88 |