Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HES7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027974932.1 | XP_027830733.1 | 2 - 3 | 426 | 90 | |
XM_027974933.1 | XP_027830734.1 | 2 - 3 | 426 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |