Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015103162.2 | XP_014958648.2 | 11 - 12 | 1434 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Ovis aries |