Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MILR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004013030.4 | XP_004013079.1 | 2 - 3 | 678 | 95 | |
XM_012186329.3 | XP_012041719.1 | 2 - 3 | 678 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |