Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGALS3BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027974210.1 | XP_027830011.1 | 3 - 4 | 413 | 66 | |
XM_027974211.1 | XP_027830012.1 | 3 - 4 | 404 | 66 |