Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NGRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017754.4 | XP_004017803.2 | 2 - 3 | 819 | 107 |
Gene Symbol: LOC114109001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027957058.1 | XP_027812859.1 | 2 - 3 | 320 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |