Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012173915.2 | XP_012029305.1 | 7 - 8 | 1321 | 84 | |
XM_012173916.3 | XP_012029306.1 | 6 - 7 | 736 | 84 | ||
XM_027967128.1 | XP_027822929.1 | 9 - 10 | 1438 | 84 | ||
XM_027967130.1 | XP_027822931.1 | 7 - 8 | 946 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |