Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027977629.1 | XP_027833430.1 | 2 - 3 | 742 | 109 | |
XM_027977630.1 | XP_027833431.1 | 3 - 4 | 765 | 109 | ||
XM_027977631.1 | XP_027833432.1 | 3 - 4 | 2418 | 109 | ||
XM_027977632.1 | XP_027833433.1 | 3 - 4 | 795 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |