Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNOC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004438.4 | XP_004004487.2 | 2 - 3 | 238 | 71 | |
XM_012128493.3 | XP_011983883.3 | 2 - 3 | 214 | 71 | ||
XM_012128504.3 | XP_011983894.2 | 2 - 3 | 234 | 71 | ||
XM_027964602.1 | XP_027820403.1 | 2 - 3 | 239 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |