Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC6A20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027957739.1 | XP_027813540.1 | 4 - 5 | 584 | 74 | |
XM_027957740.1 | XP_027813541.1 | 4 - 5 | 584 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |