Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027960705.1 | XP_027816506.1 | 3 - 4 | 398 | 64 | |
XM_027960706.1 | XP_027816507.1 | 2 - 3 | 213 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |