Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC45A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027972827.1 | XP_027828628.1 | 5 - 6 | 2317 | 57 | |
XM_027972829.1 | XP_027828630.1 | 5 - 6 | 2315 | 57 | ||
XM_027972830.1 | XP_027828631.1 | 5 - 6 | 2319 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |