Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CREG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002732.4 | XP_004002781.1 | 2 - 3 | 525 | 108 | |
XM_027977835.1 | XP_027833636.1 | 2 - 3 | 483 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |