Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP4B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012178868.2 | XP_012034258.2 | 9 - 10 | 1684 | 60 | |
XM_012178872.3 | XP_012034262.2 | 9 - 10 | 1676 | 60 | ||
XM_027973323.1 | XP_027829124.1 | 9 - 10 | 1588 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |