Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004015334.4 | XP_004015383.1 | 8 - 9 | 1066 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |