Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002559.4 | XP_004002608.2 | 2 - 3 | 1407 | 70 | |
XM_027975753.1 | XP_027831554.1 | 2 - 3 | 1407 | 70 | ||
XM_027975761.1 | XP_027831562.1 | 2 - 3 | 791 | 70 | ||
XM_027975769.1 | XP_027831570.1 | 2 - 3 | 664 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |