Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC10A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027971954.1 | XP_027827755.1 | 1 - 2 | 3268 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |