Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPSL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012097064.3 | XP_011952454.1 | 4 - 5 | 672 | 96 | |
XM_027980119.1 | XP_027835920.1 | 4 - 5 | 673 | 96 | ||
XM_027980120.1 | XP_027835921.1 | 4 - 5 | 674 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |