Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004020842.4 | XP_004020891.1 | 1 - 2 | 817 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |