Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027961293.1 | XP_027817094.1 | 4 - 5 | 988 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |