Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRUNE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027974596.1 | XP_027830397.1 | 6 - 7 | 879 | 87 | |
XM_027974599.1 | XP_027830400.1 | 6 - 7 | 879 | 87 | ||
XM_027974606.1 | XP_027830407.1 | 2 - 3 | 168 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |