Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MNAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004010698.4 | XP_004010747.1 | 1 - 2 | 221 | 83 | |
XM_012181783.3 | XP_012037173.1 | 1 - 2 | 221 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |