Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017713.4 | XP_004017762.1 | 3 - 4 | 670 | 86 | |
XM_012098328.3 | XP_011953718.1 | 3 - 4 | 948 | 86 | ||
XM_012098331.3 | XP_011953721.1 | 3 - 4 | 927 | 86 | ||
XM_012098332.2 | XP_011953722.1 | 3 - 4 | 637 | 86 | ||
XM_012098338.3 | XP_011953728.3 | 3 - 4 | 462 | 86 | ||
XM_015101856.2 | XP_014957342.1 | 3 - 4 | 755 | 86 | ||
XM_015101857.2 | XP_014957343.1 | 3 - 4 | 461 | 86 | ||
XM_027957001.1 | XP_027812802.1 | 2 - 3 | 1424 | 86 | ||
XM_027957002.1 | XP_027812803.1 | 3 - 4 | 2394 | 86 | ||
XM_027957003.1 | XP_027812804.1 | 3 - 4 | 2179 | 86 | ||
XM_027957004.1 | XP_027812805.1 | 2 - 3 | 1443 | 86 |