Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027971681.1 | XP_027827482.1 | 3 - 4 | 1955 | 63 | |
XM_027971682.1 | XP_027827483.1 | 3 - 4 | 1880 | 63 | ||
XM_027971683.1 | XP_027827484.1 | 3 - 4 | 1952 | 63 | ||
XM_027971684.1 | XP_027827485.1 | 3 - 4 | 1954 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |