Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975976.1 | XP_027831777.1 | 3 - 4 | 1546 | 102 | |
XM_027975977.1 | XP_027831778.1 | 3 - 4 | 421 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |