Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRGQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015100037.2 | XP_014955523.2 | 2 - 3 | 615 | 59 | |
XM_027978418.1 | XP_027834219.1 | 2 - 3 | 663 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |