Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015094219.2 | XP_014949705.2 | 7 - 8 | 865 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |