Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAFAH1B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012095505.3 | XP_011950895.1 | 5 - 6 | 515 | 113 | |
XM_027979229.1 | XP_027835030.1 | 5 - 6 | 536 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |