Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027974180.1 | XP_027829981.1 | 8 - 9 | 1433 | 64 | |
XM_027974181.1 | XP_027829982.1 | 8 - 9 | 1481 | 64 | ||
XM_027974182.1 | XP_027829983.1 | 7 - 8 | 1603 | 64 | ||
XM_027974183.1 | XP_027829984.1 | 8 - 9 | 1385 | 64 | ||
XM_027974184.1 | XP_027829985.1 | 8 - 9 | 1435 | 64 | ||
XM_027974185.1 | XP_027829986.1 | 8 - 9 | 1433 | 64 | ||
XM_027974186.1 | XP_027829987.1 | 8 - 9 | 1431 | 64 | ||
XM_027974187.1 | XP_027829988.1 | 8 - 9 | 1433 | 64 |