Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP298 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002768.3 | XP_004002817.2 | 4 - 5 | 1267 | 96 | |
XM_027978283.1 | XP_027834084.1 | 2 - 3 | 233 | 96 |