Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPE65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002070.4 | XP_004002119.1 | 10 - 11 | 1240 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |