Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLRG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027968864.1 | XP_027824665.1 | 5 - 6 | 1711 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |