Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZACN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004013114.4 | XP_004013163.2 | 2 - 3 | 1620 | 62 | |
XM_027974111.1 | XP_027829912.1 | 2 - 3 | 1620 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |