Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZACN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004013114.4 | XP_004013163.2 | 3 - 4 | 1664 | 94 | |
XM_027974111.1 | XP_027829912.1 | 3 - 4 | 1664 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |