Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZIC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959518.1 | XP_027815319.1 | 1 - 2 | 175 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |