Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRE11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012095281.2 | XP_011950671.2 | 12 - 13 | 1543 | 106 | |
XM_012095282.3 | XP_011950672.2 | 12 - 13 | 1507 | 106 | ||
XM_012095283.2 | XP_011950673.2 | 10 - 11 | 1169 | 106 | ||
XM_012095284.2 | XP_011950674.2 | 9 - 10 | 1097 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |