Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004020693.4 | XP_004020742.2 | 1 - 2 | 95 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |