Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002385.4 | XP_004002434.2 | 1 - 2 | 694 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |