Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002190.3 | XP_004002239.1 | 4 - 5 | 591 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |