Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012187050.3 | XP_012042440.2 | 2 - 3 | 493 | 99 |
Gene Symbol: LOC114117329 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027976157.1 | XP_027831958.1 | 2 - 3 | 499 | 99 |