Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCTS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004022355.4 | XP_004022404.1 | 3 - 4 | 598 | 77 | |
XM_004022356.4 | XP_004022405.1 | 3 - 4 | 455 | 77 | ||
XM_012107356.3 | XP_011962746.1 | 3 - 4 | 497 | 77 | ||
XM_027962993.1 | XP_027818794.1 | 3 - 4 | 638 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |