Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004009623.4 | XP_004009672.1 | 2 - 3 | 441 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Ovis aries |