Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004003247.4 | XP_004003296.2 | 3 - 4 | 943 | 109 | |
XM_012101344.3 | XP_011956734.2 | 4 - 5 | 737 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |