Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMPRSS11F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027970749.1 | XP_027826550.1 | 2 - 3 | 366 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |