Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012877.4 | XP_004012926.2 | 2 - 3 | 531 | 62 | |
GenBank mRNA | HQ283084.1 | - | 2 - 3 | 530 | 62 |