Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015097742.2 | XP_014953228.2 | 3 - 4 | 713 | 100 | |
XM_027972914.1 | XP_027828715.1 | 4 - 5 | 796 | 100 |