Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAS1R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004014026.4 | XP_004014075.1 | 1 - 2 | 196 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Ovis aries |