Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012106414.3 | XP_011961804.1 | 5 - 6 | 1267 | 88 | |
XM_012106416.3 | XP_011961806.1 | 5 - 6 | 1007 | 88 | ||
XM_015104833.2 | XP_014960319.1 | 5 - 6 | 1236 | 88 | ||
XM_015104836.2 | XP_014960322.1 | 5 - 6 | 1237 | 88 | ||
XM_027963307.1 | XP_027819108.1 | 5 - 6 | 1630 | 88 | ||
XM_027963308.1 | XP_027819109.1 | 5 - 6 | 1168 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |